missing translation for 'onlineSavingsMsg'
Learn More

Acacetin 98.0+%, TCI America™

Catalog No. A301325MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A301325MG 25 mg
A3013100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A301325MG Supplier TCI America Supplier No. A301325MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 480-44-4
Molecular Formula C16H12O5
Molecular Weight (g/mol) 284.267
MDL Number MFCD00016936
InChI Key DANYIYRPLHHOCZ-UHFFFAOYSA-N
Synonym acacetin, linarigenin, 5,7-dihydroxy-4'-methoxyflavone, acacetine, 4'-methoxyapigenin, buddleoflavonol, linarisenin, akatsetin, apigenin 4'-methyl ether, 5,7-dihydroxy-2-4-methoxyphenyl-4h-chromen-4-one
PubChem CID 5280442
ChEBI CHEBI:15335
IUPAC Name 5,7-dihydroxy-2-(4-methoxyphenyl)chromen-4-one
SMILES COC1=CC=C(C=C1)C2=CC(=O)C3=C(C=C(C=C3O2)O)O

Specifications

Melting Point 269°C
Color White-Yellow
Quantity 25 mg
Formula Weight 284.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Acacetin
RTECSNumber DJ3002000
TSCA No
Recommended Storage Refrigerator