missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acacetin

Catalog No. AC465232500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465232500 250 mg
AC465230010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465232500 Supplier Thermo Scientific Chemicals Supplier No. 465232500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 480-44-4 | C16H12O5 | 284.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 480-44-4
Molecular Formula C16H12O5
Molecular Weight (g/mol) 284.27
InChI Key DANYIYRPLHHOCZ-UHFFFAOYSA-N
IUPAC Name 5,7-dihydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one
SMILES COC1=CC=C(C=C1)C1=CC(=O)C2=C(O)C=C(O)C=C2O1

Specifications

Melting Point 260.0°C to 265.0°C
Quantity 250 mg
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in dmso, hot alcohol and alkalies, practically insoluble in ether
Formula Weight 284.26
Chemical Name or Material Acacetin
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Suspected of causing genetic defects.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

EINECSNumber 207-552-3

RUO – Research Use Only