accessibility menu, dialog, popup

UserName

OHOHC

CAS RN 471-25-0

IUPAC Name: prop-2-ynoic acid
Synonyms: propiolic acid acetylenic acid propinsaeure HC#ccooh monosodium salt propiolic acid propiolsaeure propinic acid
Molecular Weight (g/mol): 70.05
Molecular Formula: C3H2O2
InChi Key: UORVCLMRJXCDCP-UHFFFAOYSA-N
SMILES: OC(=O)C#C
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)