missing translation for 'onlineSavingsMsg'
Learn More

Propiolic Acid 97.0+%, TCI America™

Catalog No. P04975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P04975G 5 g
P049725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P04975G Supplier TCI America Supplier No. P04975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 471-25-0
Molecular Formula C3H2O2
Molecular Weight (g/mol) 70.05
MDL Number MFCD00004360
InChI Key UORVCLMRJXCDCP-UHFFFAOYSA-N
Synonym propiolic acid, 2-propynoic acid, propynoic acid, propargylic acid, acetylenecarboxylic acid, carboxyacetylene, propiolic acidd, propiolicacid, unii-p2qw39g9lz, propynoate
PubChem CID 10110
ChEBI CHEBI:33199
IUPAC Name prop-2-ynoic acid
SMILES OC(=O)C#C

Specifications

Melting Point 9°C
Color Yellow
Boiling Point 58°C
Quantity 5 g
UN Number 2920
Formula Weight 70.05
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Propiolic Acid
EINECSNumber (2)-1086
RTECSNumber UD9300000
TSCA Yes