missing translation for 'onlineSavingsMsg'
Learn More

Propiolic acid, 98+%

Catalog No. AAA1324522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1324522 100 g
AAA1324506 5 g
AAA1324514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1324522 Supplier Thermo Scientific Chemicals Supplier No. A1324522

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 471-25-0 | C3H2O2 | 70.05 g/mol

Propiolic acid is a reagent used in the preparation of transition metal complexes, haloalkyl propiolates and halopropenoates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propiolic acid is a reagent used in the preparation of transition metal complexes, haloalkyl propiolates and halopropenoates.

Solubility
Miscible with water, ether, ethanol and chloroform.

Notes
Store in cool place. Incompatible with bases, oxidizing agents and reducing agents.

Chemical Identifiers

CAS 471-25-0
Molecular Formula C3H2O2
Molecular Weight (g/mol) 70.05
MDL Number MFCD00004360
InChI Key UORVCLMRJXCDCP-UHFFFAOYSA-N
Synonym propiolic acid, 2-propynoic acid, propynoic acid, propargylic acid, acetylenecarboxylic acid, carboxyacetylene, propiolic acidd, propiolicacid, unii-p2qw39g9lz, propynoate
PubChem CID 10110
ChEBI CHEBI:33199
IUPAC Name prop-2-ynoic acid
SMILES OC(=O)C#C

Specifications

Melting Point 18°C
Density 1.136
Boiling Point 102°C (200 mmHg)
Flash Point 58°C (136°F)
Refractive Index 1.433
Quantity 100 g
UN Number UN2920
Beilstein 878176
Merck Index 14,7824
Solubility Information Miscible with water,ether,ethanol and chloroform.
Formula Weight 70.05
Percent Purity ≥98%
Chemical Name or Material Propiolic acid
Show More Show Less
Hazard Category H226-H301+H311-H314
Hazard Statement GHS H Statement
H301-H310-H314-H318-H226
Toxic if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 207-437-8
RTECSNumber UD9300000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only