accessibility menu, dialog, popup

UserName

IFFF

CAS RN 460-37-7

IUPAC Name: 1,1,1-trifluoro-3-iodopropane
Synonyms: 1,1,1-trifluoro-3-iodopropane
Molecular Weight (g/mol): 223.97
Molecular Formula: C3H4F3I
InChi Key: ULIYQAUQKZDZOX-UHFFFAOYSA-N
SMILES: FC(F)(F)CCI
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (4)