missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-3-iodopropane 98.0+%, TCI America™

Catalog No. T25885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25885G 5 g
T258825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25885G Supplier TCI America Supplier No. T25885G

Chemical Identifiers

CAS 460-37-7
Molecular Formula C3H4F3I
Molecular Weight (g/mol) 223.965
MDL Number MFCD00038531
InChI Key ULIYQAUQKZDZOX-UHFFFAOYSA-N
Synonym 1-iodo-3,3,3-trifluoropropane, 3,3,3-trifluoropropyl iodide, 3-iodo-1,1,1-trifluoropropane, 1-iodo-3,3,3-trifluoro propane, 3,3,3-trifluoropropyliodide, 1-iodo-2-perfluoroalkyl ethane, 1,1,1-trifluoro-3-iodo-propane, propane, 1,1,1-trifluoro-3-iodo, trifluoroiodopropane
PubChem CID 156165
IUPAC Name 1,1,1-trifluoro-3-iodopropane
SMILES C(CI)C(F)(F)F

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 223.96
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1-Trifluoro-3-iodopropane