missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-3,3,3-trifluoropropane, 98%

Catalog No. AC397740050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397740050 5 g Glass bottle
AC397740010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397740050 Supplier Thermo Scientific Chemicals Supplier No. 397740050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 460-37-7 | C3H4F3I | 223.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 460-37-7
Molecular Formula C3H4F3I
Molecular Weight (g/mol) 223.96
MDL Number MFCD00038531
InChI Key ULIYQAUQKZDZOX-UHFFFAOYSA-N
Synonym 1-iodo-3,3,3-trifluoropropane, 3,3,3-trifluoropropyl iodide, 3-iodo-1,1,1-trifluoropropane, 1-iodo-3,3,3-trifluoro propane, 3,3,3-trifluoropropyliodide, 1-iodo-2-perfluoroalkyl ethane, 1,1,1-trifluoro-3-iodo-propane, propane, 1,1,1-trifluoro-3-iodo, trifluoroiodopropane
PubChem CID 156165
IUPAC Name 1,1,1-trifluoro-3-iodopropane
SMILES C(CI)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Pink
Density 1.9110g/mL
Boiling Point 89.0°C to 91.0°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4190 to 1.4210 (20°C, 589nm)
Linear Formula I(CH2)2CF3
Quantity 5 g
Specific Gravity 1.911
Formula Weight 223.96
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Iodo-3, 3, 3-trifluoropropane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only