accessibility menu, dialog, popup

UserName

CAS RN 4584-49-0

IUPAC Name: hydrogen (2-chloropropyl)dimethylamine chloride
Synonyms: hydrogen (2-chloropropyl)dimethylamine chloride
Molecular Weight (g/mol): 158.07
Molecular Formula: C5H13Cl2N
InChi Key: OCWGRWAYARCRTQ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(Cl)CN(C)C
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)