missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminoisopropyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. AAB2088236
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2088236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAB2088236 Supplier Thermo Scientific Chemicals Supplier No. B2088236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 4584-49-0 | Molecular Formula: C5H13Cl2N | Molecular Weight: 158.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4584-49-0
Molecular Formula C5H13Cl2N
Molecular Weight (g/mol) 158.066
MDL Number MFCD00012534
InChI Key OCWGRWAYARCRTQ-UHFFFAOYSA-N
Synonym 2-dimethylaminoisopropyl chloride hydrochloride, 2-chloro-1-dimethylamino propane hydrochloride, 2-chloro-n,n-dimethylpropan-1-amine hydrochloride, n,n-dimethyl-2-chloropropylamine hydrochloride, 2-chloropropyldimethylammonium chloride, 2-dimethylamino isopropyl chloride hydrochloride, 2-chloropropyldimethylamine hydrochloride, 2-chloropropyl-dimethylamine hydrochloride, 1-methyl-2-dimethylaminoethyl chloride hydrochloride, 2-chloropropyl dimethylamine hydrochloride
PubChem CID 94294
IUPAC Name 2-chloro-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CC(CN(C)C)Cl.Cl

Specifications

Melting Point 190°C to 193°C
Quantity 500 g
Beilstein 3670215
Sensitivity Hygroscopic
Formula Weight 158.07
Percent Purity 98%
Chemical Name or Material 2-Dimethylaminoisopropyl chloride hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 224-971-7
RTECSNumber TY1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only