missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-(dimethylamino)propane Hydrochloride 98.0+%, TCI America™

Catalog No. D1665500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1665500G 500 g
D166525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1665500G Supplier TCI America Supplier No. D1665500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4584-49-0
Molecular Formula C5H13Cl2N
Molecular Weight (g/mol) 158.066
MDL Number MFCD00012534
InChI Key OCWGRWAYARCRTQ-UHFFFAOYSA-N
Synonym 2-dimethylaminoisopropyl chloride hydrochloride, 2-chloro-1-dimethylamino propane hydrochloride, 2-chloro-n,n-dimethylpropan-1-amine hydrochloride, n,n-dimethyl-2-chloropropylamine hydrochloride, 2-chloropropyldimethylammonium chloride, 2-dimethylamino isopropyl chloride hydrochloride, 2-chloropropyldimethylamine hydrochloride, 2-chloropropyl-dimethylamine hydrochloride, 1-methyl-2-dimethylaminoethyl chloride hydrochloride, 2-chloropropyl dimethylamine hydrochloride
PubChem CID 94294
IUPAC Name 2-chloro-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CC(CN(C)C)Cl.Cl

Specifications

Melting Point 189°C
Color White-Yellow
Quantity 500 g
Formula Weight 158.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-1-(dimethylamino)propane Hydrochloride
RTECSNumber TY1225000
TSCA Yes