CAS RN 458-37-7
Curcumin, Certified Reference Material, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.
Curcumin, 95% (total curcuminoid content), from Turmeric rhizome
CAS: 458-37-7 Molecular Formula: C21H20O6 Molecular Weight (g/mol): 368.39 MDL Number: MFCD00008365 InChI Key: ZIUSSTSXXLLKKK-KOBPDPAPSA-N Synonym: curcumin,diferuloylmethane,natural yellow 3,turmeric yellow,turmeric,curcuma,kacha haldi,gelbwurz,halad,curcumin i PubChem CID: 969516 ChEBI: CHEBI:3962 IUPAC Name: (1E,6E)-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione SMILES: COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O
Thermo Scientific Chemicals Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%
CAS: 458-37-7 Molecular Formula: C21H20O6 Molecular Weight (g/mol): 368.39 MDL Number: MFCD00008365 InChI Key: ZIUSSTSXXLLKKK-KOBPDPAPSA-N Synonym: curcumin,diferuloylmethane,natural yellow 3,turmeric yellow,turmeric,curcuma,kacha haldi,gelbwurz,halad,curcumin i PubChem CID: 969516 ChEBI: CHEBI:3962 SMILES: COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O
Curcumin (Natural), TCI America™
CAS: 458-37-7 Molecular Formula: C21H20O6 Molecular Weight (g/mol): 368.39 MDL Number: MFCD00008365 InChI Key: ZIUSSTSXXLLKKK-KOBPDPAPSA-N Synonym: curcumin,diferuloylmethane,natural yellow 3,turmeric yellow,turmeric,curcuma,kacha haldi,gelbwurz,halad,curcumin i PubChem CID: 969516 ChEBI: CHEBI:3962 IUPAC Name: (1E,4Z,6E)-5-hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,4,6-trien-3-one SMILES: COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O
Curcumin (Synthetic) 97.0+%, TCI America™
CAS: 458-37-7 Molecular Formula: C21H20O6 Molecular Weight (g/mol): 368.39 MDL Number: MFCD00008365 InChI Key: ZIUSSTSXXLLKKK-KOBPDPAPSA-N Synonym: curcumin,diferuloylmethane,natural yellow 3,turmeric yellow,turmeric,curcuma,kacha haldi,gelbwurz,halad,curcumin i PubChem CID: 969516 ChEBI: CHEBI:3962 IUPAC Name: (1E,4Z,6E)-5-hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,4,6-trien-3-one SMILES: COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O
Curcumin (0.1% in ca. 95% Ethanol), TCI America™
CAS: 458-37-7 Molecular Formula: C21H20O6 Molecular Weight (g/mol): 368.39 MDL Number: MFCD00008365 InChI Key: ZIUSSTSXXLLKKK-KOBPDPAPSA-N Synonym: curcumin,diferuloylmethane,natural yellow 3,turmeric yellow,turmeric,curcuma,kacha haldi,gelbwurz,halad,curcumin i PubChem CID: 969516 ChEBI: CHEBI:3962 IUPAC Name: (1E,4Z,6E)-5-hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,4,6-trien-3-one SMILES: COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O