missing translation for 'onlineSavingsMsg'
Learn More

Curcumin, Spectrum™ Chemical

Catalog No. 18610084
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610084 500 g Amber Glass Bottle
18610080 10 g Amber Glass Bottle
18610082 25 g Amber Glass Bottle
18610081 100 g Amber Glass Bottle
18610083 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610084 Supplier Spectrum Chemical Mfg Cor Supplier No. C1452500GMBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 458-37-7

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 g
Molecular Weight (g/mol) 368.39
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Curcumin

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.