missing translation for 'onlineSavingsMsg'
Learn More

Curcumin, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023014
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023014 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023014 Supplier Merck Emd Millipore Supplier No. PHR220950MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 458-37-7
Molecular Formula C21H20O6
Linear Formula [HOC6H3(OCH3)CH=CHCO]2 CH2
MDL Number MFCD00008365
Quantity 50 mg
Synonym Curcumin; (E,E)-1,7-bis(4-Hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione; Diferuloylmethane; Diferulylmethane; Natural Yellow 3
Vapor Density 13 (vs air)
Formula Weight 368.38
Grade Certified Reference Material