missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 96%

Catalog No. AC218582500
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218582500 250 g Glass bottle
AC218580100 10 g Glass bottle
AC218580500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC218582500 Supplier Thermo Scientific Chemicals Supplier No. 218582500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 458-37-7 | C21H20O6 | 368.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 458-37-7
Molecular Formula C21H20O6
Molecular Weight (g/mol) 368.39
MDL Number MFCD00008365
InChI Key ZIUSSTSXXLLKKK-KOBPDPAPSA-N
Synonym curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i
PubChem CID 969516
ChEBI CHEBI:3962
SMILES COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O

Specifications

Melting Point 183.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Refractive Index 1.4155 to 1.4175
Linear Formula [HOC6H3(OCH3)CH=CHCO]2CH2
Quantity 250 g
Beilstein 08,IV,3697
Merck Index 15,2663
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in diethyl ether,soluble in alcohol,glacial acetic acid,acetone
IUPAC Name (1E,6E)-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
Formula Weight 368.38
Percent Purity 98+%
Physical Form Fine Crystalline Powder
Chemical Name or Material Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
EINECSNumber 207-280-5

RUO – Research Use Only