accessibility menu, dialog, popup

UserName

H3C(S)HN(S)(R)CH3(R)(S)(S)(S)OH(R)H3C(S)H3CO(R)

CAS RN 4449-51-8

IUPAC Name: (3S,3'R,3'aS,6'S,6aS,6bS,7'aR,9R,11aS,11bR)-3',6',10,11b-tetramethyl-1,2,3,3'a,4,4',5',6,6',6a,6b,7,7',7'a,8,11,11a,11b-octadecahydro-3'H-spiro[cyclohexa[a]fluorene-9,2'-furo[3,2-b]pyridin]-3-ol
Synonyms: cyclopamine 11-deoxyjervine 11-deoxojervine 11-deoxo- (7CI) jervine jervine, 11-deoxo- (7CI)
Molecular Weight (g/mol): 411.63
Molecular Formula: C27H41NO2
InChi Key: QASFUMOKHFSJGL-LAFRSMQTSA-N
SMILES: C[C@@H]1[C@@H]2NC[C@@H](C)C[C@H]2O[C@]11CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC2=C1C
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results