missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopamine, 99%
Description
Cyclopamine, is used as a hedgehog [Hh] signaling pathway and Smo inhibitor. It depletes stem-like cancer cells in glioblastoma and blocks tumor engraftment. Can also be used to induce differentiation of human embryonic stem cells (hESCs) into hormone-expressing endocrine cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclopamine, is used as a hedgehog [Hh] signaling pathway and Smo inhibitor. It depletes stem-like cancer cells in glioblastoma and blocks tumor engraftment. Can also be used to induce differentiation of human embryonic stem cells (hESCs) into hormone-expressing endocrine cells.
Solubility
Soluble in ethanol (5 mg/ml warm), DMSO (4 mg/ml), DMF (10 mg/ml), methylene chloride, chloroform, methanol (7 mg/ml), and benzene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
Cyclopamine, is used as a hedgehog [Hh] signaling pathway and Smo inhibitor. It depletes stem-like cancer cells in glioblastoma and blocks tumor engraftment. Can also be used to induce differentiation of human embryonic stem cells (hESCs) into hormone-expressing endocrine cells.
Solubility
Soluble in ethanol (5 mg/ml warm), DMSO (4 mg/ml), DMF (10 mg/ml), methylene chloride, chloroform, methanol (7 mg/ml), and benzene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 234°C to 237°C |
| Color | White |
| Quantity | 50 mg |
| Solubility Information | Soluble in ethanol (5mg/ml warm),DMSO (4mg/ml),DMF (10mg/ml),methylene chloride,chloroform,methanol (7mg/ml),and benzene. |
| IUPAC Name | (3S,3'R,3'aS,6'S,6aS,6bS,7'aR,9R,11aS,11bR)-3',6',10,11b-tetramethyl-1,2,3,3'a,4,4',5',6,6',6a,6b,7,7',7'a,8,11,11a,11b-octadecahydro-3'H-spiro[cyclohexa[a]fluorene-9,2'-furo[3,2-b]pyridin]-3-ol |
| Formula Weight | 411.62 |
| Percent Purity | ≥99% |
| Physical Form | Powder |
| Chemical Name or Material | Cyclopamine |
Safety and Handling
| Hazard Category | H341-H361 |
| Hazard Statement | GHS H Statement H341-H361 Suspected of causing genetic defects. Suspected of damaging fertility or the unborn child. |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| RTECSNumber | GY0750000 |
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only