accessibility menu, dialog, popup

UserName

BH3COHHO

CAS RN 4433-63-0

IUPAC Name: ethylboronic acid
Synonyms: ethylboronic acid
Molecular Weight (g/mol): 73.89
Molecular Formula: C2H7BO2
InChi Key: PAVZHTXVORCEHP-UHFFFAOYSA-N
SMILES: CCB(O)O
Quantity 
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)