missing translation for 'onlineSavingsMsg'
Learn More

Ethylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. E09131G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E09131G Supplier TCI America Supplier No. E09131G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4433-63-0
Molecular Formula C2H7BO2
Molecular Weight (g/mol) 73.89
MDL Number MFCD01074536
InChI Key PAVZHTXVORCEHP-UHFFFAOYSA-N
PubChem CID 521157
IUPAC Name ethylboronic acid
SMILES CCB(O)O

Specifications

Color White-Yellow
Quantity 1 g
Synonym boronic acid, ethyl, ethaneboronic acid, ethyldihydroxyborane, ethyl boronic acid, boronic acid, ethyl-9ci, ethylboronicacid, pubchem7960, ethylboric acid, etb oh 2
Formula Weight 73.89
Physical Form Crystalline Powder
Chemical Name or Material Ethylboronic Acid (contains varying amounts of Anhydride)