missing translation for 'onlineSavingsMsg'
Learn More

Ethylboronic acid, 97%

Catalog No. AAL1995914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1995914 25 g
AAL1995903 1 g
AAL1995906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1995914 Supplier Thermo Scientific Chemicals Supplier No. L1995914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4433-63-0 | C2H7BO2 | 73.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4433-63-0
Molecular Formula C2H7BO2
Molecular Weight (g/mol) 73.89
MDL Number MFCD01074536
InChI Key PAVZHTXVORCEHP-UHFFFAOYSA-N
Synonym boronic acid, ethyl, ethaneboronic acid, ethyldihydroxyborane, ethyl boronic acid, boronic acid, ethyl-9ci, ethylboronicacid, pubchem7960, ethylboric acid, etb oh 2
PubChem CID 521157
IUPAC Name ethylboronic acid
SMILES CCB(O)O

Specifications

Melting Point 84°C to 88°C
Odor Odorless
Linear Formula CH3CH2B(OH)2
Quantity 25 g
Beilstein 1731546
Sensitivity Air sensitive
Formula Weight 73.89
Percent Purity 98%
Chemical Name or Material Ethylboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only