accessibility menu, dialog, popup

UserName

CAS RN 4253-89-8

IUPAC Name: 2-(propan-2-yldisulfanyl)propane
Synonyms: isopropyl disulfide
Molecular Weight (g/mol): 150.3
Molecular Formula: C6H14S2
InChi Key: LZAZXBXPKRULLB-UHFFFAOYSA-N
SMILES: CC(C)SSC(C)C
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)