missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl disulfide, 96%

Catalog No. AC122700250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122700250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC122700250 Supplier Thermo Scientific Chemicals Supplier No. 122700250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4253-89-8 | C6H14S2 | 150.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4253-89-8
Molecular Formula C6H14S2
Molecular Weight (g/mol) 150.3
MDL Number MFCD00008894
InChI Key LZAZXBXPKRULLB-UHFFFAOYSA-N
Synonym diisopropyl disulfide, isopropyl disulfide, diisopropyl disulphide, disulfide, bis 1-methylethyl, 2,2'-dithiodipropane, 1,2-diisopropyldisulfane, 2,5-dimethyl-3,4-dithiahexane, isopropyldisulfide, unii-bp550p623a, isopropyl disulfide 8ci
PubChem CID 77932
IUPAC Name 2-(propan-2-yldisulfanyl)propane
SMILES CC(C)SSC(C)C

Specifications

Color Beige to White
Density 0.9430g/mL
Boiling Point 175°C to 176°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4900 to 1.492
Linear Formula [(CH3)2CH]2S2
Quantity 25 g
Beilstein 01, 367
Specific Gravity 0.943
Formula Weight 150.3
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Isopropyl disulfide, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-225-

RUO – Research Use Only