missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Disulfide 98.0+%, TCI America™

Catalog No. D258225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D258225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D258225G Supplier TCI America Supplier No. D258225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4253-89-8
Molecular Formula C6H14S2
Molecular Weight (g/mol) 150.298
MDL Number MFCD00008894
InChI Key LZAZXBXPKRULLB-UHFFFAOYSA-N
Synonym diisopropyl disulfide, isopropyl disulfide, diisopropyl disulphide, disulfide, bis 1-methylethyl, 2,2'-dithiodipropane, 1,2-diisopropyldisulfane, 2,5-dimethyl-3,4-dithiahexane, isopropyldisulfide, unii-bp550p623a, isopropyl disulfide 8ci
PubChem CID 77932
IUPAC Name 2-(propan-2-yldisulfanyl)propane
SMILES CC(C)SSC(C)C

Specifications

Color Yellow
Boiling Point 177°C
Quantity 25 g
UN Number 1993
Formula Weight 150.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisopropyl Disulfide
EINECSNumber (2)-0477&(2)-2421
TSCA No