accessibility menu, dialog, popup

UserName

CAS RN 42454-06-8

IUPAC Name: 5-hydroxy-2-nitrobenzaldehyde
Synonyms: 5-hydroxy-2-nitrobenzaldehyde
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: XLYPHUGUKGMURE-UHFFFAOYSA-N
SMILES: OC1=CC=C(C(C=O)=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)