missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-nitrobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog No. AAL1205106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1205106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1205106 Supplier Thermo Scientific Chemicals Supplier No. L1205106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42454-06-8 | Molecular Formula: C7H5NO4 | Molecular Weight: 167.12 g/mol

5-Hydroxy-2-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hydroxy-2-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with air, oxidizing agents.

Chemical Identifiers

CAS 42454-06-8
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007332
InChI Key XLYPHUGUKGMURE-UHFFFAOYSA-N
Synonym 3-formyl-4-nitrophenol, 6-nitro-3-hydroxybenzaldehyde, 2-nitro-5-hydroxybenzaldehyde, 5-hydroxy-2-nitro-benzaldehyde, benzaldehyde, 5-hydroxy-2-nitro, unii-va926z93if, 6-nitro-3-hydroxy benzaldhyde, pubchem8205, acmc-1an3l, 4-08-00-00250 beilstein handbook reference
PubChem CID 39211
IUPAC Name 5-hydroxy-2-nitrobenzaldehyde
SMILES OC1=CC=C(C(C=O)=C1)[N+]([O-])=O

Specifications

Melting Point 166°C to 168°C
Quantity 5 g
Beilstein 1910196
Sensitivity Air Sensitive
Solubility Information Soluble in methanol.
Formula Weight 167.12
Percent Purity 97%
Chemical Name or Material 5-Hydroxy-2-nitrobenzaldehyde
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 255-832-9
RTECSNumber CU6700000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only