accessibility menu, dialog, popup

UserName

CAS RN 4180-23-8

IUPAC Name: 1-methoxy-4-[(1E)-prop-1-en-1-yl]benzene
Synonyms: anethole anethole (substance) epa pesticide chemical code 015604 anethole, (Z)-isomer fema number 2086 acintene O p-anethole anethole, (e)-isomer (z)-anethole anethole (product)
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: RUVINXPYWBROJD-ONEGZZNKSA-N
SMILES: COC1=CC=C(\C=C\C)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)