missing translation for 'onlineSavingsMsg'
Learn More

trans-Anethole, 98+%

Catalog No. AAA1348236
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1348236 500 g
AAA1348214 25 g
AAA1348222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1348236 Supplier Thermo Scientific Chemicals Supplier No. A1348236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4180-23-8 | C10H12O | 148.205 g/mol

trans-Anethole is used to inhibits lung and forestomach carcinogenesis, used as carbon and energy supplement in the culture media of Pseudomonas putida strain. It is also used as used as a flavoring substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Anethole is used to inhibits lung and forestomach carcinogenesis, used as carbon and energy supplement in the culture media of Pseudomonas putida strain. It is also used as used as a flavoring substance.

Solubility
Soluble in benzene, ethyl acetate, acetone, carbon disulfide.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 4180-23-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00009284
InChI Key RUVINXPYWBROJD-ONEGZZNKSA-N
Synonym anethole, trans-anethole, e-anethole, 4-propenylanisole, anise camphor, trans-anethol, isoestragole, e-1-methoxy-4-prop-1-en-1-yl benzene, anethol, p-anethole
PubChem CID 637563
ChEBI CHEBI:35616
IUPAC Name 1-methoxy-4-[(E)-prop-1-enyl]benzene
SMILES CC=CC1=CC=C(C=C1)OC

Specifications

Melting Point 20°C to 23°C
Density 0.991
Boiling Point 232°C to 235°C
Flash Point 96°C (205°F)
Refractive Index 1.56
Quantity 500 g
Beilstein 774229
Sensitivity Light sensitive
Merck Index 14,643
Solubility Information Soluble in benzene,ethyl acetate,acetone,carbon disulfide.
Formula Weight 148.21
Percent Purity ≥98%
Chemical Name or Material trans-Anethole, stabilized
Show More Show Less
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 224-052-0
RTECSNumber BZ9275000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only