missing translation for 'onlineSavingsMsg'
Learn More

trans-Anethole 98.0+%, TCI America™

Catalog No. P0494500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0494500G 500 g
P049425G 25 g
P0494100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0494500G Supplier TCI America Supplier No. P0494500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4180-23-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00009284
InChI Key RUVINXPYWBROJD-ONEGZZNKSA-N
Synonym anethole, trans-anethole, e-anethole, 4-propenylanisole, anise camphor, trans-anethol, isoestragole, e-1-methoxy-4-prop-1-en-1-yl benzene, anethol, p-anethole
PubChem CID 637563
ChEBI CHEBI:35616
IUPAC Name 1-methoxy-4-[(E)-prop-1-enyl]benzene
SMILES CC=CC1=CC=C(C=C1)OC

Specifications

Color Yellow
Boiling Point 235°C
Quantity 500 g
Formula Weight 148.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-Anethole
EINECSNumber (3)-0572&(3)-2762
RTECSNumber BZ9275000
TSCA Yes