missing translation for 'onlineSavingsMsg'
Learn More

trans-Anethole Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014165
Change view
Click to view available options
Quantity:
2.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014165 2.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014165 Supplier Merck Emd Millipore Supplier No. PHR12182.2ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

trans-Anethole is an aromatic monoterpenoid compound and derivative of phenylpropene. It is predominantly used as a fragrance in cosmetics and as a flavoring agent in food and pharmaceutical industries.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 4180-23-8
Density 0.988 g/mL (at 25°C)
Flash Point 91°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula CH3 CH=CHC6H4 OCH3
MDL Number MFCD00009284
Melting Point 20°C to 21°C
Boiling Point 234°C to 237°C
Synonym 4-Propenylanisole; trans-1-Methoxy-4-(1-propenyl)benzene
Type Certified Reference Material
Quantity 2.2 mL
Formula Weight 148.20
Grade Certified Reference Material
Show More Show Less