accessibility menu, dialog, popup

UserName

CH3S(S)HNOOH3CH3CCH3OONOO

CAS RN 3845-64-5

IUPAC Name: 2,5-dioxopyrrolidin-1-yl (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl (2S)-2-[(tert-butoxycarbonyl)amino]-4-(methylsulfanyl)butanoate|BOC-MET-OSU
Molecular Weight (g/mol): 346.4
Molecular Formula: C14H22N2O6S
InChi Key: PCZJWSPKNYONIM-VIFPVBQESA-N
SMILES: CSCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)