missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-methionine N-Succinimidyl Ester 97.0+%, TCI America™

Catalog No. B34335G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34335G Supplier TCI America Supplier No. B34335G

Chemical Identifiers

CAS 3845-64-5
Molecular Formula C14H22N2O6S
Molecular Weight (g/mol) 346.40
MDL Number MFCD00042760
InChI Key PCZJWSPKNYONIM-VIFPVBQESA-N
Synonym boc-met-osu, 2,5-dioxopyrrolidin-1-yl 2s-2-tert-butoxycarbonyl amino-4-methylsulfanyl butanoate, tbm-nhs, boc-l-methionine hydeoxysuccinimide ester, n-boc-l-methionine n-succinimidyl ester, n-tert-butoxycarbonyl-l-methionine n-succinimidyl ester, n-alpha-t-butyloxycarbonyl-l-methionine succinimidyl ester, 2,5-dioxo-1-pyrrolidinyl n-2-methyl-2-propanyl oxy carbonyl methioninate
PubChem CID 12876462
IUPAC Name 2,5-dioxopyrrolidin-1-yl (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoate
SMILES CSCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 126°C
Color White
Quantity 5 g
Formula Weight 346.40
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-methionine N-Succinimidyl Ester
TSCA No
Recommended Storage Freezer