missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-methionine N-succinimidyl ester, 97%

Catalog No. AAH6656314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6656314 25 g
AAH6656306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6656314 Supplier Thermo Scientific Chemicals Supplier No. H6656314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3845-64-5 | C14H22N2O6S | 346.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3845-64-5
Molecular Formula C14H22N2O6S
Molecular Weight (g/mol) 346.40
MDL Number MFCD00042760
InChI Key PCZJWSPKNYONIM-VIFPVBQESA-N
Synonym boc-met-osu, 2,5-dioxopyrrolidin-1-yl 2s-2-tert-butoxycarbonyl amino-4-methylsulfanyl butanoate, tbm-nhs, boc-l-methionine hydeoxysuccinimide ester, n-boc-l-methionine n-succinimidyl ester, n-tert-butoxycarbonyl-l-methionine n-succinimidyl ester, n-alpha-t-butyloxycarbonyl-l-methionine succinimidyl ester, 2,5-dioxo-1-pyrrolidinyl n-2-methyl-2-propanyl oxy carbonyl methioninate
PubChem CID 12876462
IUPAC Name 2,5-dioxopyrrolidin-1-yl (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoate
SMILES CSCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 120°C to 128°C
Beilstein 4238574
Quantity 25 g
Formula Weight 346.4
Sensitivity Moisture sensitive
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Boc-L-methionine N-succinimidyl ester
EINECSNumber 223-341-9
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only