accessibility menu, dialog, popup

UserName

H3COOBOHHO

CAS RN 380430-53-5

IUPAC Name: [2-(ethoxycarbonyl)phenyl]boronic acid
Synonyms: 2-(ethoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol): 193.99
Molecular Formula: C9H11BO4
InChi Key: QZKVVOXAEBCLPZ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1B(O)O
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)