missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethoxycarbonyl)benzeneboronic acid, 98%

Catalog No. AAH2586706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2586706 5 g
AAH2586703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2586706 Supplier Thermo Scientific Chemicals Supplier No. H2586706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 380430-53-5 | C9H11BO4 | 193.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 380430-53-5
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.99
MDL Number MFCD02179453
InChI Key QZKVVOXAEBCLPZ-UHFFFAOYSA-N
Synonym 2-ethoxycarbonyl phenyl boronic acid, 2-ethoxycarbonylbenzeneboronic acid, 2-ethoxycarbonyl phenylboronic acid, 2-ethoxycarbonylphenyl boronic acid, o-ethoxycarbonylphenylboronic acid, 2-carbethoxyphenylboronic acid, ethyl 2-dihydroxyboranyl benzoate, ethyl 2-boronobenzoate, 2-ethoxycarbonyl benzeneboronic acid
PubChem CID 2773405
IUPAC Name (2-ethoxycarbonylphenyl)boronic acid
SMILES CCOC(=O)C1=CC=CC=C1B(O)O

Specifications

Melting Point 130°C to 133°C
Quantity 5 g
Formula Weight 193.99
Percent Purity 98%
Chemical Name or Material 2-(Ethoxycarbonyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only