missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. E08771G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E08771G Supplier TCI America Supplier No. E08771G

Chemical Identifiers

CAS 380430-53-5
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.99
MDL Number MFCD02179453
InChI Key QZKVVOXAEBCLPZ-UHFFFAOYSA-N
PubChem CID 2773405
IUPAC Name [2-(ethoxycarbonyl)phenyl]boronic acid
SMILES CCOC(=O)C1=CC=CC=C1B(O)O

Specifications

Melting Point 138°C
Quantity 1 g
Synonym 2-ethoxycarbonyl phenyl boronic acid, 2-ethoxycarbonylbenzeneboronic acid, 2-ethoxycarbonyl phenylboronic acid, 2-ethoxycarbonylphenyl boronic acid, o-ethoxycarbonylphenylboronic acid, 2-carbethoxyphenylboronic acid, ethyl 2-dihydroxyboranyl benzoate, ethyl 2-boronobenzoate, 2-ethoxycarbonyl benzeneboronic acid
Formula Weight 193.99
Physical Form Crystalline Powder
Chemical Name or Material 2-(Ethoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride)