accessibility menu, dialog, popup

UserName

CAS RN 3724-43-4

IUPAC Name: chloro(dimethylamino)methylium chloride
Synonyms: chloro(dimethylamino)methylium chloride
Molecular Weight (g/mol): 128
Molecular Formula: C3H7Cl2N
InChi Key: QQVDYSUDFZZPSU-UHFFFAOYSA-M
SMILES: [Cl-].CN(C)[CH+]Cl
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (4)