missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethylene)dimethylammonium chloride, 96%

Catalog No. AC295770250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC295770250 25 g Glass bottle
AC295770050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC295770250 Supplier Thermo Scientific Chemicals Supplier No. 295770250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3724-43-4 | C3H7Cl2N | 128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent

Chemical Identifiers

CAS 3724-43-4
Molecular Formula C3H7Cl2N
Molecular Weight (g/mol) 128
MDL Number MFCD00011868
InChI Key QQVDYSUDFZZPSU-UHFFFAOYSA-M
Synonym arnold's reagent, vilsmeier reagent, chloromethylene dimethyliminium chloride, methanaminium, n-chloromethylene-n-methyl-, chloride, chloromethylene dimethylammonium chloride, n-chloromethylidene-n-methylmethanaminium chloride, chloromethylenedimethylammonium chloride, chloromethylidene dimethylazanium chloride, n-chloromethylene-n,n-dimethyl ammonium chloride
PubChem CID 77311
IUPAC Name chloromethylidene(dimethyl)azanium;chloride
SMILES C[N+](=CCl)C.[Cl-]

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 139°C
Color Brown to Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (Argentometry)
Packaging Glass bottle
Linear Formula ClCH=N(CH3)2Cl
Quantity 25 g
Fieser 04,186; 05,251; 06,220; 07,422; 08,186; 09,514; 10,457; 12,564; 13,341
Solubility Information Solubility in water: reacts. Other solubilities: 28g/100mL chloroform,slightly soluble in other solvents
Formula Weight 128
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material (Chloromethylene)dimethylammonium chloride, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May damage the unborn child.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
TSCA TSCA

RUO – Research Use Only