accessibility menu, dialog, popup

UserName

CAS RN 3678-63-5

IUPAC Name: 4-chloro-2-methylpyridine
Synonyms: 4-chloro-2-methylpyridine
Molecular Weight (g/mol): 127.57
Molecular Formula: C6H6ClN
InChi Key: DAOZBJCTEPJGES-UHFFFAOYSA-N
SMILES: CC1=NC=CC(Cl)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)