missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylpyridine 98.0+%, TCI America™

Catalog No. M22845G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22845G 5 g
M22841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22845G Supplier TCI America Supplier No. M22845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3678-63-5 | C6H6ClN | 127.571 g/mol

Chemical Identifiers

CAS 3678-63-5
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.571
MDL Number MFCD00038833
InChI Key DAOZBJCTEPJGES-UHFFFAOYSA-N
Synonym 4-chloro-2-picoline, 2-methyl-4-chloropyridine, 4-chloro-2-methyl-pyridine, pyridine, 4-chloro-2-methyl, p-chloropicoline, 4-chloropicoline, 4-chloro-2-picolin, acmc-20amln, pubchem2071
PubChem CID 581392
IUPAC Name 4-chloro-2-methylpyridine
SMILES CC1=NC=CC(=C1)Cl

Specifications

Color Yellow
Boiling Point 56°C
Quantity 5 g
Formula Weight 127.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-methylpyridine