missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylpyridine, 95%

Catalog No. AAH6409306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6409306 5 g
AAH6409303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6409306 Supplier Thermo Scientific Chemicals Supplier No. H6409306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3678-63-5 | C6H6ClN | 127.571 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3678-63-5
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.571
MDL Number MFCD00038833
InChI Key DAOZBJCTEPJGES-UHFFFAOYSA-N
Synonym 4-chloro-2-picoline, 2-methyl-4-chloropyridine, 4-chloro-2-methyl-pyridine, pyridine, 4-chloro-2-methyl, p-chloropicoline, 4-chloropicoline, 4-chloro-2-picolin, acmc-20amln, pubchem2071
PubChem CID 581392
IUPAC Name 4-chloro-2-methylpyridine
SMILES CC1=NC=CC(=C1)Cl

Specifications

Color Colorless to Yellow
Density 1.15 g/mL
Boiling Point 54°C to 56°C (15 mmHg)(literature)
Refractive Index 1.5205
Quantity 5 g
Beilstein 107196
Formula Weight 127.57
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-methylpyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only