CAS RN 364782-34-3
IUPAC Name:
[(1R)-1-(naphthalen-1-yl)ethyl]({3-[3-(trifluoromethyl)phenyl]propyl})amine hydrochloride
Synonyms:
sensipar cinacalcet hydrochloride cnc-hcl
Molecular Weight (g/mol):
393.88
Molecular Formula:
C22H23ClF3N
InChi Key:
QANQWUQOEJZMLL-PKLMIRHRSA-N
SMILES:
Cl.C[C@@H](NCCCC1=CC=CC(=C1)C(F)(F)F)C1=C2C=CC=CC2=CC=C1
1
–
2
of
2
results
1
Thermo Scientific™ Cinacalcet hydrochloride
CAS: 364782-34-3 Chemical Name or Material: Cinacalcet hydrochloride
2
R&D Systems™ Cinacalcet hydrochloride
A Tocris product, part of R&D Systems. Low molecular weight, organic compound that may help regulate biological process.