missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cinacalcet hydrochloride

Catalog No. 617050
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
617050 50 mg
617010 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 617050 Supplier R&D Systems™ Supplier No. 6170/50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Low molecular weight, organic compound that may help regulate biological process.

Cinacalcet hydrochloride is a calcium-sensing receptor (CaSR) allosteric agonist. Activates CaSR signaling in cellular proliferation and phosphatidylinositol (PI) hydrolysis assays. Reduces serum parathyroid hormone levels in rats. Also potent CYP2D2 inhibitor (IC50 = 87 nM) and L-type calcium channel blocker. Orally bioavailable.

Specifications

CAS 364782-34-3
Quantity 50 mg
Product Line Tocris
Target Calcium-Sensing Receptor
Molecular Formula C22H22F3N.HCl
Formula Weight 393.87g/mol
Product Type Inhibitor

For Research Use Only