missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cinacalcet hydrochloride

Catalog No. p-7214787
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466010010 1 g
AC466010050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466010010 Supplier Thermo Scientific™ Supplier No. 466010010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 364782-34-3
Chemical Name or Material Cinacalcet hydrochloride

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: slightly. Other solubilities: soluble in methanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only