accessibility menu, dialog, popup

UserName

CAS RN 364-98-7

IUPAC Name: 7-chloro-3-methyl-4H-1λ⁶,2,4-benzothiadiazine-1,1-dione
Synonyms: hyperstat proglicem mutabase proglycem hypertonalum dizoxide
Molecular Weight (g/mol): 230.67
Molecular Formula: C8H7ClN2O2S
InChi Key: GDLBFKVLRPITMI-UHFFFAOYSA-N
SMILES: CC1=NS(=O)(=O)C2=CC(Cl)=CC=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results