missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Diazoxide, 98%

Catalog No. AAJ66010ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66010ME 500 mg
AAJ66010MC 100 mg
AAJ6601003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ66010ME Supplier Thermo Scientific Chemicals Supplier No. J66010ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 364-98-7 | C8H7ClN2O2S, C8H7ClN2O2S | 230.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 364-98-7

Specifications

Quantity 500 mg
Description White to off-white
Content And Storage 2°C to 8°C
Molecular Formula C8H7ClN2O2S
Formula Weight 230.67
Synonym 7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide;
Melting Point 330°C to 331°C
Solubility Soluble in 0.1M NaOH. Insoluble in water or in methanol.
Form Powder
Assay 98%

RUO – Research Use Only