accessibility menu, dialog, popup

UserName

CAS RN 36322-90-4

IUPAC Name: (3E)-3-{hydroxy[(pyridin-2-yl)amino]methylidene}-2-methyl-3,4-dihydro-2H-1λ⁶,2-benzothiazine-1,1,4-trione
Synonyms: piroxicam
Molecular Weight (g/mol): 331.35
Molecular Formula: C15H13N3O4S
InChi Key: LDJVNWFKNMYYNT-FYWRMAATSA-N
SMILES: CN1\C(=C(\O)NC2=CC=CC=N2)C(=O)C2=CC=CC=C2S1(=O)=O
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)