missing translation for 'onlineSavingsMsg'
Learn More

Piroxicam 98.0+%, TCI America™

Catalog No. P190510G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P190510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P190510G Supplier TCI America Supplier No. P190510G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 36322-90-4
Melting Point 201°C
Color White-Yellow
Molecular Formula C15H13N3O4S
Quantity 10 g
MDL Number MFCD00057317
UN Number 2811
Synonym piroxicam, feldene, piroxicamum, pyroxycam, roxicam, piroftal, baxo, artroxicam, bruxicam, caliment
InChI Key QYSPLQLAKJAUJT-UHFFFAOYSA-N
SMILES CN1C(=C(C2=CC=CC=C2S1(=O)=O)O)C(=O)NC3=CC=CC=N3
IUPAC Name 4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-yl-1$l^{6},2-benzothiazine-3-carboxamide
Molecular Weight (g/mol) 331.346
PubChem CID 54676228
ChEBI CHEBI:8249
Formula Weight 331.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Piroxicam
Show More Show Less
RTECSNumber DL0705000
TSCA No
Recommended Storage Refrigerator