missing translation for 'onlineSavingsMsg'
Learn More

Piroxicam Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018961
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018961 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018961 Supplier Merck Emd Millipore Supplier No. PHR1588200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 36322-90-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C15H13N3O4 S
MDL Number MFCD00057317
Synonym Piroxicam; 4-Hydroxy-2-methyl-3-(pyrid-2-yl-carbamoyl)-2 H-1,2-benzothiazine 1,1-dioxide
Type Certified Reference Material
Quantity 200 mg
Formula Weight 331.35
Grade Certified Reference Material