accessibility menu, dialog, popup

UserName

CAS RN 363-81-5

IUPAC Name: 2,4,6-trifluoroaniline
Synonyms: 2,4,6-trifluoroaniline
Molecular Weight (g/mol): 147.1
Molecular Formula: C6H4F3N
InChi Key: BJSVKBGQDHUBHZ-UHFFFAOYSA-N
SMILES: NC1=C(F)C=C(F)C=C1F
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)