missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluoroaniline 98.0+%, TCI America™

Catalog No. T10385G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T10385G Supplier TCI America Supplier No. T10385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 363-81-5
Molecular Formula C6H4F3N
Molecular Weight (g/mol) 147.1
MDL Number MFCD00007650
InChI Key BJSVKBGQDHUBHZ-UHFFFAOYSA-N
Synonym 2-amino-1,3,5-trifluorobenzene, 2,4,6-trifluorobenzenamine, benzenamine, 2,4,6-trifluoro, 2,4,6-trifluorophenylamine, pubchem2468, acmc-1cmhu, 2,4,6 trifluoroaniline, 2,4,6-tri-fluoroaniline, 2,4,6-trifluoro aniline, 2,4,6-trifluoro-aniline
PubChem CID 67765
IUPAC Name 2,4,6-trifluoroaniline
SMILES C1=C(C=C(C(=C1F)N)F)F

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1325
Formula Weight 147.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4,6-Trifluoroaniline